Assay Details
Target Gene Details
Entrez Gene ID: | 29781 |
Gene Name: | non-SMC condensin II complex subunit H2 |
Gene Aliases: |
CAPH2 |
Location: |
Chr.22:50508216-50524780 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCAPH2 | NM_001185011.1 | 20 | 2842 | NP_001171940.1 |
NM_152299.3 | 20 | 2839 | NP_689512.2 |
Target Gene Details
Entrez Gene ID: | 9997 |
Gene Name: | SCO2 cytochrome c oxidase assembly protein |
Gene Aliases: |
CEMCOX1, MYP6, SCO1L |
Location: |
Chr.22:50523568-50526439 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCO2 | NM_001169109.1 | 2 | 345 | NP_001162580.1 |
NM_001169110.1 | 2 | 296 | NP_001162581.1 | |
NM_001169111.1 | 2 | 314 | NP_001162582.1 | |
NM_005138.2 | 2 | 285 | NP_005129.2 | |
AL021683.1 | 2 | 223 | CAA16671.1 | |
BC102024.1 | 2 | 213 | AAI02025.1 | |
BC102025.1 | 2 | 213 | AAI02026.1 | |
BG392981.1 | 2 | 297 | ||
BM787182.1 | 2 | 345 | ||
BQ878572.1 | 2 | 285 | ||
CN409692.1 | 2 | 253 | ||
CR456569.1 | 1 | 143 | CAG30455.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758852 | Chr.22:50491090 - 50578979 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160782 | Chr.22:50463961 - 50571524 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1057656 | Chr.22:50478783 - 50595520 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834238 | Chr.22:50402690 - 50556965 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv471224 | Chr.22:50435189 - 50580482 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv829340 | Chr.22:50505720 - 50603696 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv955201 | Chr.22:50421672 - 50537371 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834239 | Chr.22:50485317 - 50630215 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv589222 | Chr.22:50491152 - 50589773 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)