Assay Details
Target Gene Details
Entrez Gene ID: | 374291 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit S7 |
Gene Aliases: |
CI-20, CI-20KD, MY017, PSST |
Location: |
Chr.19:1383571-1395589 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFS7 | NM_024407.4 | 8 | 649 | NP_077718.3 |
AF060512.1 | 8 | 619 | AAG43130.1 | |
AF115969.1 | 8 | 632 | AAP97240.1 | |
AK056061.1 | 1 | 2054 | ||
AK091024.1 | 6 | 2918 | ||
AK091046.1 | 1 | 3611 | ||
AK091623.1 | 7 | 2689 | ||
AK092169.1 | 6 | 2957 | BAC03820.1 | |
AK127759.1 | 7 | 3737 | BAC87117.1 | |
AK128713.1 | 5 | 3224 | BAC87587.1 | |
AK222738.1 | 8 | 626 | BAD96458.1 | |
BC001715.3 | 7 | 592 | AAH01715.2 | |
BC005954.1 | 8 | 626 | AAH05954.1 | |
BC111517.1 | 8 | 636 | AAI11518.1 | |
BX099945.1 | 1 | 151 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | ADAT3 MEX3D TCF3 REEP6 CSNK1G2-AS1 C19orf25 MIR1909 PLK5 REXO1 BTBD2 APC2 GAMT DAZAP1 SCAMP4 KLF16 ONECUT3 UQCR11 RPS15 PCSK4 CSNK1G2 LOC100288123 NDUFS7 ABHD17A ADAMTSL5 ATP8B3 MBD3 MUM1 |
nsv833705 | Chr.19:1380125 - 1544934 on Build GRCh38 | Loss | APC2 GAMT NDUFS7 DAZAP1 ADAMTSL5 REEP6 C19orf25 RPS15 PLK5 PCSK4 |
nsv482207 | Chr.19:1383884 - 1395589 on Build GRCh38 | Gain | NDUFS7 |
esv3643389 | Chr.19:1264454 - 1397379 on Build GRCh38 | Gain | GAMT NDUFS7 C19orf24 EFNA2 CIRBP MUM1 CIRBP-AS1 |
esv2758743 | Chr.19:951641 - 1544934 on Build GRCh38 | Gain+Loss | ARID3A LOC105372235 C19orf24 GRIN3B REEP6 STK11 C19orf25 PLK5 EFNA2 SBNO2 LOC102725180 CIRBP-AS1 APC2 GAMT DAZAP1 MIDN POLR2E WDR18 TMEM259 RPS15 GPX4 PCSK4 ABCA7 ARHGAP45 NDUFS7 ADAMTSL5 CNN2 CBARP RNU6-2 ATP5D CIRBP MUM1 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | ADAT3 MEX3D TCF3 REEP6 CSNK1G2-AS1 C19orf25 MIR1909 PLK5 REXO1 BTBD2 APC2 GAMT DAZAP1 SCAMP4 MKNK2 KLF16 ONECUT3 UQCR11 RPS15 PCSK4 CSNK1G2 LOC100288123 NDUFS7 ABHD17A ADAMTSL5 ATP8B3 MBD3 MUM1 |
esv3643396 | Chr.19:1356856 - 1396468 on Build GRCh38 | Gain | NDUFS7 MUM1 |
nsv578278 | Chr.19:1371355 - 1448181 on Build GRCh38 | Loss | APC2 GAMT NDUFS7 DAZAP1 RPS15 MUM1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |