Assay Details
Target Gene Details
Entrez Gene ID: | 21672 |
Gene Name: | peroxiredoxin 2 |
Gene Aliases: |
AL022839, Band-8, NkefB, PRP, PrxII, TDX1, TPx, TPx-B, TR, TSA, Tdpx1, Torin |
Location: |
Chr.8:84969648-84974317 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prdx2 | NM_011563.5 | NP_035693.3 | ||
AK005225.1 | BAB23893.1 | |||
AK008433.1 | BAB25666.1 | |||
AK010653.1 | BAB27093.1 | |||
AK014765.1 | ||||
AK088280.1 | BAC40255.1 | |||
AK202198.1 | ||||
AK217812.1 | ||||
BC002034.1 | AAH02034.1 | |||
BC081454.1 | AAH81454.1 | |||
BC086783.1 | AAH86783.1 | |||
U20611.1 | AAA69475.1 | |||
U51679.1 | AAB01941.1 | |||
X82067.1 | CAA57566.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic Validated |