Assay Details
Target Gene Details
Entrez Gene ID: | 15205 |
Gene Name: | hairy and enhancer of split 1 (Drosophila) |
Gene Aliases: |
Hry, bHLHb39 |
Location: |
Chr.16:30065357-30067796 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hes1 | NM_008235.2 | NP_032261.1 | ||
AK132536.1 | BAE21226.1 | |||
AK133538.1 | BAE21713.1 | |||
BC018375.1 | AAH18375.1 | |||
BC051428.1 | AAH51428.1 |
More Information
Set Membership: |
Intragenic Non-exonic Validated |