Assay Details
Target Gene Details
Entrez Gene ID: | 16336 |
Gene Name: | insulin-like 3 |
Gene Aliases: |
Rlf, Rlnl |
Location: |
Chr.8:71689252-71690577 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Insl3 | NM_013564.7 | NP_038592.3 | ||
AK132857.1 | BAE21395.1 | |||
BC049540.1 | AAH49540.1 | |||
S82815.1 | AAB39365.1 |
Target Gene Details
Entrez Gene ID: | 16453 |
Gene Name: | Janus kinase 3 |
Gene Aliases: |
fae, wil |
Location: |
Chr.8:71676383-71690577 on Build GRCm38 |
Assay Gene Location: | Within Intron 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Jak3 | NM_010589.6 | NP_034719.2 | ||
AK038268.1 | BAC29956.1 | |||
AK088365.1 | BAC40305.1 | |||
BC105577.1 | ||||
BC131646.1 | ||||
BC131647.1 | ||||
L32955.1 | AAA21565.1 | |||
L33768.1 | AAA21415.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic Validated |