Assay Details
Target Gene Details
Entrez Gene ID: | 22377 |
Gene Name: | WW domain binding protein 1 |
Gene Aliases: |
AI255152 |
Location: |
Chr.6:83119044-83121663 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wbp1 | NM_001083922.1 | 5 | 947 | NP_001077391.1 |
NM_001083923.1 | 4 | 845 | NP_001077392.1 | |
NM_016757.2 | 5 | 950 | NP_058037.2 | |
AK076948.1 | 5 | 882 | BAC36533.1 | |
AK152822.1 | 5 | 950 | ||
AK170907.1 | 3 | 1548 | ||
AK187136.1 | 1 | 48 | ||
AK190643.1 | 1 | 46 | ||
AK197680.1 | ||||
BC005666.1 | 5 | 1066 | AAH05666.1 | |
BC019889.1 | 4 | 769 | AAH19889.1 | |
U40825.1 | 5 | 912 | AAB40892.1 |