Assay Details
Target Gene Details
Entrez Gene ID: | 66290 |
Gene Name: | ATPase, H+ transporting, lysosomal V1 subunit G1 |
Gene Aliases: |
1810024D14Rik, AA960677, ATP6J, Atp6g1, VAG1, Vma10 |
Location: |
Chr.4:63544765-63550701 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp6v1g1 | NM_024173.2 | NP_077135.1 | ||
AB076406.1 | BAC10289.1 | |||
AK003563.1 | BAB22859.1 | |||
AK007596.1 | BAB25127.1 | |||
AK010131.1 | BAB26720.1 | |||
AK081611.1 | BAC38271.1 | |||
AK088712.1 | BAC40520.1 | |||
AK152699.1 | BAE31429.1 | |||
AK159558.1 | BAE35183.1 | |||
AK168972.1 | BAE40775.1 | |||
AK169079.1 | ||||
BC003429.1 | AAH03429.1 | |||
BC088836.1 | AAH88836.1 | |||
CN838691.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic Validated |