Assay Details
Target Gene Details
Entrez Gene ID: | 16593 |
Gene Name: | kinesin light chain 1 |
Gene Aliases: |
AI874768, Kns2 |
Location: |
Chr.12:111758849-111807844 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klc1 | NM_001025358.2 | NP_001020529.2 | ||
NM_001025359.2 | NP_001020530.2 | |||
NM_001025360.2 | NP_001020531.2 | |||
NM_001025361.2 | NP_001020532.2 | |||
NM_001025362.2 | NP_001020533.2 | |||
NM_001025363.2 | NP_001020534.2 | |||
NM_001081959.1 | NP_001075428.1 | |||
NM_008450.2 | NP_032476.2 | |||
AF055665.1 | AAC27740.1 | |||
AK031309.1 | BAC27341.1 | |||
AY753300.1 | AAV30550.1 | |||
BC055744.1 | AAH55744.1 | |||
BG094009.1 | ||||
BU922789.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic Validated |