Assay Details
Target Gene Details
Entrez Gene ID: | 53357 |
Gene Name: | phospholipase A2, group VI |
Gene Aliases: |
BB112799, PNPLA9, iPLA(2)beta, iPLA2, iPLA2beta |
Location: |
Chr.15:79286228-79328530 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g6 | NM_001199023.1 | NP_001185952.1 | ||
NM_001199024.1 | NP_001185953.1 | |||
NM_001199025.1 | NP_001185954.1 | |||
NM_016915.4 | NP_058611.1 | |||
AF259401.1 | AAF72651.1 | |||
AK144495.1 | BAE25917.1 | |||
AK153333.1 | BAE31912.1 | |||
AK177026.1 | ||||
AK177982.1 | ||||
BC003487.1 | AAH03487.1 | |||
BC052845.1 | AAH52845.1 | |||
BC057209.1 | AAH57209.1 | |||
DV647961.1 | ||||
U88624.2 | AAB48511.2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic Validated |