Assay Details
Target Gene Details
Entrez Gene ID: | 268697 |
Gene Name: | cyclin B1 |
Gene Aliases: |
Ccnb1-rs1, Ccnb1-rs13, CycB1, Cycb-4, Cycb-5, Cycb1-rs1 |
Location: |
Chr.13:100778732-100786543 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccnb1 | NM_172301.3 | NP_758505.2 | ||
AK136259.1 | BAE22901.1 | |||
AK143597.1 | BAE25456.1 | |||
AK163259.1 | BAE37263.1 | |||
AK166686.1 | BAE38945.1 | |||
AK167950.1 | ||||
AK168441.1 | BAE40346.1 | |||
AK168502.1 | BAE40387.1 | |||
AK168993.1 | BAE40792.1 | |||
BC011478.1 | AAH11478.1 | |||
BC080202.1 | AAH80202.1 | |||
BC085238.1 | AAH85238.1 | |||
FJ931532.1 | ||||
S43105.1 | AAB22970.1 | |||
X58708.1 | CAA41545.1 | |||
X64713.1 | CAA45968.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic Validated |