Assay Details
Target Gene Details
Entrez Gene ID: | 676914 |
Gene Name: | arylacetamide deacetylase-like 2 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.3:59952309-59973594 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm9696 | NR_037189.1 | 5 | 1382 | |
AK140981.1 | 5 | 1385 | BAE24536.1 |
More Information
Set Membership: |
Intragenic Exonic |