Assay Details
Target Gene Details
Entrez Gene ID: | 71207 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 4 |
Gene Aliases: |
4933436C10Rik, DIPP2, DIPP2alpha, DIPP2beta, HDCMB47P |
Location: |
Chr.10:95547007-95564167 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nudt4 | NM_027722.4 | 1 | 136 | NP_081998.3 |
AK017075.1 | BAB30582.1 | |||
AK048062.1 | 1 | 116 | BAC33229.1 | |
CJ169842.1 |
More Information
Set Membership: |
Intragenic Exonic |