Assay Details
Target Gene Details
Entrez Gene ID: | 22290 |
Gene Name: | ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome |
Gene Aliases: |
Hydb, mKIAA4057 |
Location: |
Chr.Y:1085187-1245754 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uty | NM_009484.2 | NP_033510.2 | ||
AF057367.1 | AAC67385.1 | |||
AK043154.1 | BAC31478.1 | |||
AK142344.1 | ||||
AK142512.1 | BAE25094.1 | |||
AK155922.1 | ||||
AK162410.1 | BAE36901.1 | |||
AK220463.1 | BAD90490.1 |
More Information
Set Membership: |
Intragenic Non-exonic |