Assay Details
Target Gene Details
Entrez Gene ID: | 666945 |
Gene Name: | predicted gene 10638 |
Gene Aliases: |
ENSMUSG00000074178 |
Location: |
Chr.8:86745699-86747060 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm10638 | NR_027829.1 | 1 | 241 | |
AK133050.1 | 1 | 242 | BAE21488.1 |
Target Gene Details
Entrez Gene ID: | 20437 |
Gene Name: | seven in absentia 1A |
Gene Aliases: |
AA982064, AI853500, Sinh1a |
Location: |
Chr.8:86723920-86746006 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Siah1a | NM_009172.2 | 1 | 68 | NP_033198.1 |
BC046317.1 | 1 | 68 | AAH46317.1 | |
Z19579.1 | CAA79630.1 |
More Information
Set Membership: |
Intragenic Exonic |