Assay Details
Target Gene Details
Entrez Gene ID: | 235674 |
Gene Name: | acetyl-Coenzyme A acyltransferase 1B |
Gene Aliases: |
- |
Location: |
Chr.9:119148043-119157093 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acaa1b | NM_146230.3 | 1 | 73 | NP_666342.1 |
AK145953.1 | BAE26781.1 | |||
AY273812.1 | 1 | 73 | AAP31669.1 | |
BC019882.1 | AAH19882.1 |
More Information
Set Membership: |
Intragenic Exonic |