Assay Details
Target Gene Details
Entrez Gene ID: | 72735 |
Gene Name: | Egfr long non-coding downstream RNA |
Gene Aliases: |
2810442I21Rik, 4930500E12Rik, Fabl |
Location: |
Chr.11:16934709-16951282 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eldr | NR_110421.1 | 1 | 176 | |
NR_110422.1 | 1 | 176 | ||
NR_110423.1 | 1 | 176 | ||
NR_110424.1 | 1 | 176 | ||
NR_110425.1 | 1 | 176 | ||
AK013290.1 | ||||
AK015657.1 | 1 | 177 | ||
AK141825.1 | ||||
AK165613.1 | ||||
BI990615.1 |