Assay Details
Target Gene Details
Entrez Gene ID: | 73139 |
Gene Name: | centromere protein V |
Gene Aliases: |
3110013H01Rik, AA671303, Prr6 |
Location: |
Chr.11:62524944-62539502 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cenpv | NM_028448.1 | 1 | 368 | NP_082724.1 |
AK014046.1 | 1 | 369 | BAB29130.1 | |
BC147794.1 | 1 | 330 | ||
BC147795.1 | 1 | 330 | ||
BC147835.1 | 1 | 330 | ||
BC147837.1 | 1 | 330 |