Assay Details
Target Gene Details
Entrez Gene ID: | 22631 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta polypeptide |
Gene Aliases: |
1110013I11Rik, 14-3-3zeta, AI596267, AL022924, AU020854 |
Location: |
Chr.15:36770261-36802984 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ywhaz | NM_001253805.1 | 6 | 2833 | NP_001240734.1 |
NM_001253806.1 | 6 | 2558 | NP_001240735.1 | |
NM_001253807.1 | 5 | 2393 | NP_001240736.1 | |
NM_011740.3 | 7 | 3030 | NP_035870.1 | |
AK146800.1 | 6 | 2549 | BAE27442.1 | |
AK149013.1 | 1 | 1069 | ||
AK151506.1 | 4 | 2029 | BAE30457.1 | |
AK151900.1 | 6 | 2522 | BAE30783.1 | |
AK167128.1 | 6 | 2530 | BAE39275.1 | |
BC050891.1 | 7 | 2736 | AAH50891.1 | |
D78647.1 | 6 | 2481 | BAA11464.1 |
More Information
Set Membership: |
Intragenic Exonic |