Assay Details
Target Gene Details
Entrez Gene ID: | 223669 |
Gene Name: | zinc finger protein 7 |
Gene Aliases: |
KRAB20, KRAB7, Krox-2, Zfp-7, Zfp65, Zfp80, Zfp86-rs1, mszf73-2 |
Location: |
Chr.15:76879217-76892395 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp7 | NM_145916.2 | 5 | 2624 | NP_666021.2 |
AK144887.1 | 5 | 2836 | BAE26117.1 | |
AK168948.1 | 5 | 2579 | BAE40754.1 | |
BC011501.1 | 5 | 2642 | AAH11501.1 |