Assay Details
Target Gene Details
Entrez Gene ID: | 66105 |
Gene Name: | ubiquitin-conjugating enzyme E2D 3 |
Gene Aliases: |
1100001F19Rik, 9430029A22Rik, AA414951 |
Location: |
Chr.3:135438244-135467180 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ube2d3 | NM_025356.4 | 8 | 2298 | NP_079632.1 |
AK049906.1 | 8 | 2299 | BAC33981.1 | |
AK077666.1 | 8 | 2358 | BAC36940.1 |
More Information
Set Membership: |
Intragenic Exonic |