Assay Details
Target Gene Details
Entrez Gene ID: | 21985 |
Gene Name: | tumor protein D52 |
Gene Aliases: |
AI043038, mD52 |
Location: |
Chr.3:8928626-9004769 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tpd52 | NM_001025261.1 | 8 | 2302 | NP_001020432.1 |
NM_001025262.1 | 6 | 2233 | NP_001020433.1 | |
NM_001025263.1 | 7 | 2178 | NP_001020434.1 | |
NM_001025264.1 | 7 | 2163 | NP_001020435.1 | |
NM_009412.2 | 6 | 2136 | NP_033438.1 | |
AK005456.1 | 6 | 2117 | BAB24048.1 | |
AK032111.1 | 7 | 2141 | BAC27709.1 | |
AK150935.1 | 6 | 2099 | ||
AK151471.1 | 6 | 2097 | BAE30428.1 | |
AK151744.1 | 6 | 2097 | BAE30655.1 | |
AK152543.1 | 6 | 2097 | BAE31298.1 | |
AK153266.1 | 6 | 2097 | BAE31856.1 | |
AK153292.1 | BAE31875.1 | |||
AK161753.1 | BAE36558.1 | |||
AK168379.1 | 6 | 2092 | BAE40308.1 | |
AK168817.1 | 6 | 2100 | BAE40644.1 | |
AK169751.1 | BAE41345.1 | |||
AK170725.1 | 6 | 2082 | BAE41982.1 | |
AK171050.1 | 6 | 2077 | BAE42212.1 | |
AY048852.1 | AAL05266.1 | |||
BC004068.1 | AAH04068.1 | |||
BC094018.1 | AAH94018.1 | |||
U44426.1 | AAB40897.1 |