Assay Details
Target Gene Details
Entrez Gene ID: | 16796 |
Gene Name: | LIM and SH3 protein 1 |
Gene Aliases: |
AA408629, Def-4, SH3P6, Tg(Col1a1-lacZ)1Ngma |
Location: |
Chr.11:97799005-97838765 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lasp1 | NM_010688.4 | 7 | 2354 | NP_034818.1 |
AK034404.1 | 7 | 2355 | BAC28696.1 | |
AK049199.1 | 7 | 2355 | BAC33604.1 | |
AK078445.1 | 7 | 2355 | BAC37278.1 | |
AK148858.1 | 7 | 2357 | BAE28681.1 | |
AK154656.1 | 7 | 2355 | BAE32746.1 | |
AK168521.1 | 7 | 2355 | BAE40400.1 | |
AK169381.1 | 7 | 2355 | BAE41128.1 | |
AK179064.1 | 1 | 120 | ||
AK181281.1 | 1 | 120 | ||
AK206302.1 | 1 | 120 | ||
AK212158.1 | 1 | 120 | ||
AK213957.1 | 1 | 120 | ||
AK214011.1 | 1 | 120 | ||
BC010840.1 | 7 | 2339 | AAH10840.1 |