Assay Details
Target Gene Details
Entrez Gene ID: | 67291 |
Gene Name: | coiled-coil domain containing 137 |
Gene Aliases: |
3110023B02Rik |
Location: |
Chr.11:120458129-120464358 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc137 | NM_152807.3 | 6 | 2089 | NP_690020.1 |
AK013412.1 | 1 | 581 | ||
AK156682.1 | 6 | 2098 | BAE33806.1 | |
BC026662.1 | 6 | 2091 | AAH26662.1 |