Assay Details
Target Gene Details
Entrez Gene ID: | 216233 |
Gene Name: | suppressor of cytokine signaling 2 |
Gene Aliases: |
8030460M17, AI527257, AW108012, CIS2, Cish2, D130043N08Rik, JAB, SOCS-2, SSI-2, hg |
Location: |
Chr.10:95383903-95417253 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Socs2 | NM_001168655.1 | 3 | 1506 | NP_001162126.1 |
NM_001168656.1 | 3 | 1464 | NP_001162127.1 | |
NM_001168657.1 | 2 | 1385 | NP_001162128.1 | |
NM_007706.4 | 3 | 1533 | NP_031732.1 | |
AK033206.1 | BAC28196.2 | |||
AK083884.1 | ||||
AK137403.1 | 3 | 1413 | BAE23343.1 | |
AK139048.1 | 1 | 329 | ||
AK144810.1 | 3 | 1475 | ||
AK162591.1 | 3 | 1498 | BAE36978.1 |
More Information
Set Membership: |
Intragenic Exonic |