Assay Details
Target Gene Details
Entrez Gene ID: | 14718 |
Gene Name: | glutamic-oxaloacetic transaminase 1, soluble |
Gene Aliases: |
AI789014, Got-1, cAspAT, cCAT |
Location: |
Chr.19:43499752-43524605 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Got1 | NM_010324.2 | 9 | 1394 | NP_034454.2 |
AK146445.1 | 9 | 1296 | BAE27177.1 | |
AK147807.1 | 9 | 1317 | ||
AK147934.1 | 9 | 1298 | ||
BC002057.1 | 9 | 1282 | AAH02057.1 | |
J02623.1 | 9 | 1298 | AAA37263.1 |
More Information
Set Membership: |
Intragenic Exonic |