Assay Details
Target Gene Details
Entrez Gene ID: | 54609 |
Gene Name: | ubiquilin 2 |
Gene Aliases: |
Chap1, Dsk2, HRIHFB2157, Plic-2, Plic2 |
Location: |
Chr.X:153498232-153501562 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ubqln2 | NM_018798.2 | 1 | 1106 | NP_061268.2 |
AF177346.1 | 1 | 1107 | AAF01366.1 | |
AK076159.1 | 1 | 1110 | ||
BC021824.1 | 1 | 491 | AAH21824.1 | |
BC053022.1 | 1 | 1106 | AAH53022.1 |