Assay Details
Target Gene Details
Entrez Gene ID: | 72065 |
Gene Name: | RAP2C, member of RAS oncogene family |
Gene Aliases: |
2010200P20Rik, AI194294, AL022976 |
Location: |
Chr.X:51003912-51020092 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rap2c | NM_172413.2 | NP_766001.1 | ||
AK087971.1 | BAC40066.1 | |||
AK146057.1 | BAE26866.1 | |||
AK146956.1 | BAE27564.1 | |||
AK147015.1 | BAE27609.1 | |||
BC027363.1 | AAH27363.3 | |||
BC050056.1 | ||||
BC064814.1 | AAH64814.2 | |||
BC094890.1 | AAH94890.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)