Assay Details
Target Gene Details
Entrez Gene ID: | 66294 |
Gene Name: | family with sequence similarity 3, member A |
Gene Aliases: |
1810037C20Rik |
Location: |
Chr.X:74384719-74393280 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam3a | NM_025473.3 | 9 | 1175 | NP_079749.1 |
AK003680.1 | 1 | 121 | ||
AK007715.1 | 9 | 1176 | BAB25208.1 | |
AK012353.1 | 5 | 503 | ||
AK214462.1 | ||||
AK219974.1 | ||||
BC069871.1 | 9 | 1310 | AAH69871.1 |
More Information
Set Membership: |
Intragenic Exonic |