Assay Details
Target Gene Details
Entrez Gene ID: | 209268 |
Gene Name: | immunoglobulin superfamily, member 1 |
Gene Aliases: |
5330413N23, 5530402E03, AI747649, InhBP/p120, mKIAA0364 |
Location: |
Chr.X:49782536-49797906 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igsf1 | NM_177591.4 | 14 | 2646 | NP_808259.2 |
NM_183336.2 | 5 | 900 | NP_899179.1 | |
AK030701.1 | 5 | 902 | ||
AK076427.1 | 5 | 893 | BAC36338.1 | |
AK129126.1 | 14 | 2628 | BAC97936.1 | |
AY227771.1 | 14 | 2640 | AAP57079.1 | |
AY227774.1 | 5 | 895 | AAP57082.1 | |
BC119264.1 | 5 | 863 | ||
BC119266.1 | 5 | 863 |
More Information
Set Membership: |
Intragenic Exonic |