Assay Details
Target Gene Details
Entrez Gene ID: | 107146 |
Gene Name: | glycine-N-acyltransferase |
Gene Aliases: |
A330009E03Rik, ACGNAT, AI195249, AI315345, CAT, GAT |
Location: |
Chr.19:12633308-12651737 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Glyat | NM_145935.3 | 1 | 93 | NP_666047.1 |
AK039262.1 | 1 | 94 | BAC30297.1 |
More Information
Set Membership: |
Intragenic Exonic |