Assay Details
Target Gene Details
Entrez Gene ID: | 14537 |
Gene Name: | glucosaminyl (N-acetyl) transferase 1, core 2 |
Gene Aliases: |
5630400D21Rik, B130048E03, C2 GlcNAcT, C2GNT, IGnT |
Location: |
Chr.19:17326141-17372844 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gcnt1 | NM_001136484.3 | 3 | 3810 | NP_001129956.3 |
NM_010265.5 | 3 | 3851 | NP_034395.4 | |
NM_173442.5 | 3 | 3883 | NP_775618.3 | |
AK017462.1 | 1 | 322 | ||
AK045216.1 | 3 | 3834 | BAC32265.1 | |
BC039126.1 | 3 | 3808 | AAH39126.1 |
More Information
Set Membership: |
Intragenic Exonic |