Assay Details
Target Gene Details
Entrez Gene ID: | 18846 |
Gene Name: | plexin A3 |
Gene Aliases: |
PlexA3, Plxa3, Plxn3, Plxn4, SEX, mKIAA4078 |
Location: |
Chr.X:74329066-74344689 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plxna3 | NM_008883.2 | 3 | 866 | NP_032909.2 |
BC093482.1 | 3 | 849 | ||
D86950.1 | 3 | 832 | BAA13190.1 |