Assay Details
Target Gene Details
Entrez Gene ID: | 18846 |
Gene Name: | plexin A3 |
Gene Aliases: |
PlexA3, Plxa3, Plxn3, Plxn4, SEX, mKIAA4078 |
Location: |
Chr.X:74329066-74344689 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plxna3 | NM_008883.2 | 23 | 4232 | NP_032909.2 |
AJ748653.1 | 5 | 712 | CAG38687.1 | |
AK049319.1 | 5 | 646 | BAC33681.1 | |
AK220285.1 | 19 | 2856 | BAD90210.1 | |
BC093482.1 | 23 | 3984 | ||
D86950.1 | 23 | 4198 | BAA13190.1 |