Assay Details
Target Gene Details
Entrez Gene ID: | 226016 |
Gene Name: | abhydrolase domain containing 17B |
Gene Aliases: |
5730446C15Rik, Cgi67, Fam108b, Fam108b1 |
Location: |
Chr.19:21653309-21685638 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abhd17b | NM_146096.3 | 2 | 514 | NP_666208.2 |
AK050879.1 | 2 | 639 |
More Information
Set Membership: |
Intragenic Exonic |