Assay Details
Target Gene Details
Entrez Gene ID: | 21873 |
Gene Name: | tight junction protein 2 |
Gene Aliases: |
ZO-2 |
Location: |
Chr.19:24094502-24225128 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tjp2 | NM_001198985.1 | 21 | 3184 | NP_001185914.1 |
NM_011597.4 | 21 | 3174 | NP_035727.2 | |
AF113005.1 | 21 | 3032 | AAD19964.1 | |
AK028494.1 | 21 | 3102 | ||
AK209802.1 | ||||
BC034677.1 | 21 | 3092 | AAH34677.1 | |
BC039924.1 | 21 | 3103 | AAH39924.1 | |
CF736959.1 | 2 | 171 |
More Information
Set Membership: |
Intragenic Exonic |