Assay Details
Target Gene Details
Entrez Gene ID: | 102902673 |
Gene Name: | predicted gene 21992 |
Gene Aliases: |
Rbm14-rbm4 |
Location: |
Chr.19:4784293-4811634 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm21992 | NM_001290127.1 | NP_001277056.1 | ||
NM_001290128.1 | NP_001277057.1 | |||
AK087488.1 | BAC39895.1 | |||
EU287940.1 | ||||
EU287941.1 |
Target Gene Details
Entrez Gene ID: | 56275 |
Gene Name: | RNA binding motif protein 14 |
Gene Aliases: |
1300007E16Rik, PSP2, Sytip, p16, p16K |
Location: |
Chr.19:4800566-4811634 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rbm14 | NM_019869.3 | 2 | 1680 | NP_063922.2 |
AK004928.1 | 2 | 1650 | BAB23678.1 | |
AK050945.1 | 2 | 1682 | ||
AK088201.1 | 2 | 1646 | BAC40206.1 | |
AK153230.1 | 3 | 1556 | BAE31823.1 | |
AK167503.1 | 2 | 1659 | BAE39579.1 | |
BC010294.1 | 2 | 1631 | AAH10294.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |