Assay Details
Target Gene Details
Entrez Gene ID: | 226278 |
Gene Name: | prolactin releasing hormone receptor |
Gene Aliases: |
Gm339, Gpr10, Gr3, Prrpr |
Location: |
Chr.19:60465641-60469431 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prlhr | NM_201615.2 | 1 | 1040 | NP_963909.2 |
BC125649.1 | 1 | 972 | ||
BC125651.1 | 1 | 972 |
More Information
Set Membership: |
Intragenic Exonic |