Assay Details
Target Gene Details
Entrez Gene ID: | 58859 |
Gene Name: | epidermal growth factor-containing fibulin-like extracellular matrix protein 2 |
Gene Aliases: |
0610011K11Rik, Fbln4, MBP1 |
Location: |
Chr.19:5473960-5481854 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Efemp2 | NM_001164352.1 | 10 | 1647 | NP_001157824.1 |
NM_021474.3 | 11 | 1406 | NP_067449.3 | |
AF104223.1 | 10 | 1361 | AAD45219.1 | |
AF109122.1 | 10 | 1620 | AAF65189.1 | |
AI327154.1 | 1 | 156 | ||
AK075585.1 | 11 | 1395 | BAC35838.1 | |
BC012269.1 | 10 | 1544 | AAH12269.1 |
More Information
Set Membership: |
Intragenic Exonic |