Assay Details
Target Gene Details
Entrez Gene ID: | 15183 |
Gene Name: | histone deacetylase 3 |
Gene Aliases: |
AW537363 |
Location: |
Chr.18:37936971-37955000 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac3 | NM_010411.2 | 15 | 1647 | NP_034541.2 |
AF074881.1 | 15 | 1594 | AAC36305.1 | |
AF074882.1 | 9 | 1021 | AAC36306.1 | |
AF098295.1 | 15 | 1572 | AAF28798.1 | |
AF125536.1 | 15 | 1579 | AAF36425.1 | |
AK145158.1 | 15 | 1613 | BAE26265.1 |