Assay Details
Target Gene Details
Entrez Gene ID: | 268977 |
Gene Name: | latent transforming growth factor beta binding protein 1 |
Gene Aliases: |
9430031G15Rik, 9830146M04, Ltbp-1, Tgfb, b2b1000Clo |
Location: |
Chr.17:75005529-75392967 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ltbp1 | NM_019919.3 | 3 | 1052 | NP_064303.2 |
AF022889.1 | 3 | 1017 | AAC33307.1 | |
CV559283.1 | 2 | 187 |
More Information
Set Membership: |
Intragenic Exonic |