Assay Details
Target Gene Details
Entrez Gene ID: | 18596 |
Gene Name: | platelet derived growth factor receptor, beta polypeptide |
Gene Aliases: |
AI528809, CD140b, PDGFR-1, Pdgfr |
Location: |
Chr.18:61045150-61085068 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdgfrb | NM_001146268.1 | 23 | 4789 | NP_001139740.1 |
NM_008809.2 | 23 | 4786 | NP_032835.2 | |
AK134785.1 | 24 | 4811 | BAE22283.1 | |
AK143247.1 | 23 | 4768 | ||
BC055311.1 | 23 | 4801 | AAH55311.1 | |
DQ979390.1 | 17 | 3442 | ||
X04367.1 | 23 | 4495 | CAA27882.1 |
More Information
Set Membership: |
Intragenic Exonic |