Assay Details
Target Gene Details
Entrez Gene ID: | 18596 |
Gene Name: | platelet derived growth factor receptor, beta polypeptide |
Gene Aliases: |
AI528809, CD140b, PDGFR-1, Pdgfr |
Location: |
Chr.18:61045150-61085068 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdgfrb | NM_001146268.1 | 1 | 209 | NP_001139740.1 |
NM_008809.2 | 1 | 209 | NP_032835.2 | |
AK048486.1 | 1 | 202 | ||
AK134785.1 | 1 | 160 | BAE22283.1 | |
AK143247.1 | 1 | 189 | ||
BC055311.1 | 1 | 221 | AAH55311.1 | |
BY753644.1 | 1 | 212 |
More Information
Set Membership: |
Intragenic Exonic |