Assay Details
Target Gene Details
Entrez Gene ID: | 208177 |
Gene Name: | pleckstrin homology-like domain, family B, member 2 |
Gene Aliases: |
AV253284, C820004H04Rik, LL5b, LL5beta |
Location: |
Chr.16:45746230-45953701 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phldb2 | NM_001252442.1 | 19 | 4802 | NP_001239371.1 |
NM_153412.4 | 18 | 4643 | NP_700461.2 | |
AF506821.1 | 18 | 4644 | AAM33635.1 | |
AK076541.1 | 1 | 375 | ||
AK087133.1 | 3 | 1031 | ||
BC050915.1 | 19 | 4658 | AAH50915.1 | |
BC060683.1 | 19 | 4658 | AAH60683.1 |