Assay Details
Target Gene Details
Entrez Gene ID: | 76498 |
Gene Name: | progestin and adipoQ receptor family member IV |
Gene Aliases: |
1500004C10Rik |
Location: |
Chr.17:23736186-23740898 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Paqr4 | NM_023824.3 | 3 | 1902 | NP_076313.1 |
AB041564.1 | 3 | 1823 | BAA95048.1 | |
AK005142.1 | 3 | 1925 | BAB23839.1 | |
AK033625.1 | 3 | 1920 | BAC28398.1 | |
AK044092.1 | 3 | 1722 | BAC31770.1 | |
AK044486.1 | BAC31947.1 | |||
AK152676.1 | 3 | 1901 | BAE31408.1 | |
AY424293.1 | 3 | 1853 | AAR08381.1 | |
BC011185.1 | 3 | 1902 | AAH11185.1 | |
BC026921.1 | 3 | 1853 | AAH26921.1 |
Target Gene Details
Entrez Gene ID: | 268930 |
Gene Name: | protein kinase, membrane associated tyrosine/threonine 1 |
Gene Aliases: |
6230424P17, AW209059, Myt1 |
Location: |
Chr.17:23726336-23736735 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pkmyt1 | NM_023058.3 | NP_075545.2 | ||
AF175892.1 | AAG09210.1 | |||
AK158550.1 | BAE34554.1 | |||
AK165164.1 | BAE38056.1 | |||
AK200332.1 | ||||
AK212231.1 | ||||
BC025061.1 | AAH25061.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |