Assay Details
Target Gene Details
Entrez Gene ID: | 100384868 |
Gene Name: | predicted gene, 37013 |
Gene Aliases: |
Pcdha4-g |
Location: |
Chr.18:36952648-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm37013 | NM_001174154.2 | NP_001167625.1 |
Target Gene Details
Entrez Gene ID: | 103611158 |
Gene Name: | predicted gene, 38666 |
Gene Aliases: |
Pcdha6-g |
Location: |
Chr.18:36967631-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm38666 | NM_001301256.1 | NP_001288185.1 |
Target Gene Details
Entrez Gene ID: | 103611159 |
Gene Name: | protocadherin alpha 7-gamma |
Gene Aliases: |
- |
Location: |
Chr.18:36973802-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdha7-g | NM_001301259.1 | NP_001288188.1 |
Target Gene Details
Entrez Gene ID: | 93709 |
Gene Name: | protocadherin gamma subfamily A, 1 |
Gene Aliases: |
- |
Location: |
Chr.18:37661945-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga1 | NM_033584.1 | NP_291062.1 | ||
AY013792.1 | AAK26081.1 |
Target Gene Details
Entrez Gene ID: | 93710 |
Gene Name: | protocadherin gamma subfamily A, 2 |
Gene Aliases: |
mKIAA0588 |
Location: |
Chr.18:37669105-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga2 | NM_033585.1 | NP_291063.1 | ||
AB093250.1 | BAC41434.1 | |||
AY013796.1 | AAK26085.1 | |||
BC139178.1 |
Target Gene Details
Entrez Gene ID: | 93711 |
Gene Name: | protocadherin gamma subfamily A, 3 |
Gene Aliases: |
- |
Location: |
Chr.18:37674335-37841873 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga3 | NM_033586.2 | NP_291064.1 | ||
AY013797.1 | AAK26086.1 | |||
BC060691.1 | AAH60691.1 |
Target Gene Details
Entrez Gene ID: | 93712 |
Gene Name: | protocadherin gamma subfamily A, 4 |
Gene Aliases: |
- |
Location: |
Chr.18:37685400-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga4 | NM_033587.3 | NP_291065.3 | ||
AY013798.1 | AAK26087.1 | |||
BC049814.1 | AAH49814.1 |
Target Gene Details
Entrez Gene ID: | 93713 |
Gene Name: | protocadherin gamma subfamily A, 5 |
Gene Aliases: |
- |
Location: |
Chr.18:37694501-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga5 | NM_033588.4 | NP_291066.1 | ||
AK043491.1 | BAC31558.1 | |||
AY013799.1 | AAK26088.1 | |||
BC057404.1 | AAH57404.1 |
Target Gene Details
Entrez Gene ID: | 93714 |
Gene Name: | protocadherin gamma subfamily A, 6 |
Gene Aliases: |
- |
Location: |
Chr.18:37707229-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga6 | NM_033589.1 | NP_291067.1 | ||
AY013800.1 | AAK26089.1 | |||
BC080852.1 | AAH80852.1 |
Target Gene Details
Entrez Gene ID: | 93715 |
Gene Name: | protocadherin gamma subfamily A, 7 |
Gene Aliases: |
- |
Location: |
Chr.18:37714834-37841873 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhga7 | NM_033590.3 | 1 | 1699 | NP_291068.2 |
AK034910.1 | 1 | 1716 | BAC28880.1 | |
AY013801.1 | 1 | 1591 | AAK26090.1 | |
BC077712.1 | 1 | 1699 | AAH77712.1 |
Target Gene Details
Entrez Gene ID: | 93699 |
Gene Name: | protocadherin gamma subfamily B, 1 |
Gene Aliases: |
- |
Location: |
Chr.18:37680458-37841870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb1 | NM_033574.3 | NP_291052.3 | ||
AY013804.1 | AAK26093.1 | |||
BC054741.1 | AAH54741.1 | |||
BC098367.1 | AAH98367.1 |
Target Gene Details
Entrez Gene ID: | 93700 |
Gene Name: | protocadherin gamma subfamily B, 2 |
Gene Aliases: |
- |
Location: |
Chr.18:37689859-37841872 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcdhgb2 | NM_033575.3 | NP_291053.1 | ||
AF464151.1 | AAM93554.1 | |||
AF464152.1 | AAM93555.1 | |||
AY013805.1 | AAK26094.1 | |||
BC054557.1 | AAH54557.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |