Assay Details
Target Gene Details
Entrez Gene ID: | 67394 |
Gene Name: | RIKEN cDNA 4930404I05 gene |
Gene Aliases: |
- |
Location: |
Chr.16:91011249-91016503 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4930404I05Rik | NR_028368.1 | |||
AK006191.1 | ||||
AK015081.1 |
Target Gene Details
Entrez Gene ID: | 104015 |
Gene Name: | synaptojanin 1 |
Gene Aliases: |
A930006D20Rik, AA675315, mKIAA0910 |
Location: |
Chr.16:90936092-91012137 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Synj1 | NM_001164483.1 | NP_001157955.1 | ||
AW045957.1 |
More Information
Set Membership: |
Intragenic Non-exonic |