Assay Details
Target Gene Details
Entrez Gene ID: | 545253 |
Gene Name: | predicted gene 5820 |
Gene Aliases: |
EG545253 |
Location: |
Chr.18:38892456-38971274 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm5820 | NM_001033789.2 | 5 | 738 | NP_001028961.1 |
AK145364.1 | 5 | 739 | BAE26390.1 | |
BC147564.1 | 6 | 689 |