Assay Details
Target Gene Details
Entrez Gene ID: | 27632 |
Gene Name: | negative elongation factor complex member E, Rdbp |
Gene Aliases: |
D17H6S45, NELF-E, RD, Rdbp |
Location: |
Chr.17:34850391-34856372 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nelfe | NM_001045863.1 | 7 | 652 | NP_001039328.1 |
NM_001045864.1 | 7 | 738 | NP_001039329.1 | |
NM_138580.2 | 7 | 526 | NP_613046.2 | |
AK011663.1 | 7 | 510 | ||
AK012389.1 | 7 | 543 | BAB28206.1 | |
AK131947.1 | 7 | 510 | BAE20893.1 | |
AK147011.1 | 7 | 634 | BAE27605.1 | |
BC034867.1 | 7 | 478 | AAH34867.1 | |
CN525174.1 | ||||
DV662511.1 | 4 | 271 |