Assay Details
Target Gene Details
Entrez Gene ID: | 52040 |
Gene Name: | protein phosphatase 1, regulatory subunit 10 |
Gene Aliases: |
2610025H06Rik, Cat53, D17Ertd808e, Fb19, Pnuts |
Location: |
Chr.17:35916512-35952996 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r10 |