Assay Details
Target Gene Details
Entrez Gene ID: | 381122 |
Gene Name: | calpain 13 |
Gene Aliases: |
CANP 13, Gm943 |
Location: |
Chr.17:73306456-73399306 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Capn13 | NM_001033444.2 | 23 | 2259 | NP_001028616.1 |
AK136569.1 | 23 | 2260 | BAE23051.1 | |
BC147510.1 | 23 | 2176 | ||
BC147512.1 | 23 | 2176 |
More Information
Set Membership: |
Intragenic Exonic |